ENA189566716 Enamine

3-[(pyridin-3-yl)methoxy]aniline

  • MDL number MFCD09046941
  • Cas Number : 105326-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43724
Availability details for
Quantity
  • Availability
  • Add to Cart