ENA189566719 Enamine

2-chloro-1-[2,5-dimethyl-1-(2-methylphenyl)-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD08444935
  • Cas Number : 923743-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43729
Availability details for
Quantity
  • Availability
  • Add to Cart