ENA189566723 Enamine

1-{3-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]phenyl}methanamine dihydrochloride

  • MDL number MFCD11858129
  • Cas Number : 1170427-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43762
Availability details for
Quantity
  • Availability
  • Add to Cart