ENA189566736 Enamine

4-(benzenesulfonyl)phenol

  • MDL number MFCD00025037
  • Cas Number : 7402-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42972
Availability details for
Quantity
  • Availability
  • Add to Cart