ENA189566737 Enamine

{2-[(2-methylphenyl)amino]-1,3-thiazol-4-yl}methanol hydrochloride

  • MDL number MFCD11858000
  • Cas Number : 1170156-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42984
Availability details for
Quantity
  • Availability
  • Add to Cart