ENA196829966 Enamine

cyano[(methylsulfanyl)(morpholin-4-yl)methylidene]amine

  • Cas Number : 55843-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40837
Availability details for
Quantity
  • Availability
  • Add to Cart