ENA196829976 Enamine

3-[(pyridin-2-yl)amino]propanoic acid hydrochloride

  • MDL number MFCD12197343
  • Cas Number : 1181458-18-3
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43882
Availability details for
Quantity
  • Availability
  • Add to Cart