ENA196829978 Enamine

4H-1,2,4-triazol-3-ylmethanamine dihydrochloride

  • MDL number MFCD11506345
  • Cas Number : 859791-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44109

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-44109
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart