ENA196830001 Enamine

6-ethyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine hydrochloride

  • MDL number MFCD12197355
  • Cas Number : 1181458-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45000
Availability details for
Quantity
  • Availability
  • Add to Cart