ENA196830002 Enamine

3-[(benzylamino)methyl]benzamide hydrochloride

  • MDL number MFCD12197141
  • Cas Number : 1181457-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-45007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45007
Availability details for
Quantity
  • Availability
  • Add to Cart