ENA196830006 Enamine

1-(3-aminopropyl)-5-bromo-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD12197145
  • Cas Number : 1181458-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45041
Availability details for
Quantity
  • Availability
  • Add to Cart