ENA196830007 Enamine

N-(5-amino-1,3,4-thiadiazol-2-yl)methanesulfonamide

  • MDL number MFCD12197148
  • Cas Number : 1100459-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45088
Availability details for
Quantity
  • Availability
  • Add to Cart