ENA196830009 Enamine

3-chloro-2-(2-methyl-1H-imidazol-1-yl)aniline dihydrochloride

  • MDL number MFCD12197159
  • Cas Number : 1181458-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45173
Availability details for
Quantity
  • Availability
  • Add to Cart