ENA196830011 Enamine

2-[(piperidin-4-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione hydrochloride

  • MDL number MFCD12197163
  • Cas Number : 124939-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45185
Availability details for
Quantity
  • Availability
  • Add to Cart