ENA196830012 Enamine

2-amino-N-[(4-chlorophenyl)methyl]-2-phenylacetamide hydrochloride

  • MDL number MFCD12197164
  • Cas Number : 1214011-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45187
Availability details for
Quantity
  • Availability
  • Add to Cart