ENA196830013 Enamine

4-({[(3-fluorophenyl)methyl]amino}methyl)benzonitrile hydrochloride

  • MDL number MFCD12197165
  • Cas Number : 1181458-00-3
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45191

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-45191
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart