ENA196830018 Enamine

2-(3-amino-2-hydroxypropyl)-2,3-dihydro-1H-isoindole-1,3-dione hydrochloride

  • MDL number MFCD12197181
  • Cas Number : 289050-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45275
Availability details for
Quantity
  • Availability
  • Add to Cart