ENA196830019 Enamine

5-[(3-chlorophenyl)methyl]-4-methyl-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD12197362
  • Cas Number : 1181458-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45298
Availability details for
Quantity
  • Availability
  • Add to Cart