ENA196830021 Enamine

4-methyl-5-[(2-methylphenyl)methyl]-1,3-thiazol-2-amine hydrobromide

  • MDL number MFCD12197186
  • Cas Number : 1181457-99-7
  • Molecular Weight : 299.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45302
Availability details for
Quantity
  • Availability
  • Add to Cart