ENA196830023 Enamine

2-amino-2-phenyl-1-(piperidin-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD12197191
  • Cas Number : 24185-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45322
Availability details for
Quantity
  • Availability
  • Add to Cart