ENA196830026 Enamine

3-amino-N-[(2E)-1-methylazepan-2-ylidene]benzene-1-sulfonamide

  • MDL number MFCD12197365
  • Cas Number : 1181459-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45382
Availability details for
Quantity
  • Availability
  • Add to Cart