ENA196830029 Enamine

methyl 2-{[(2-chlorophenyl)methyl]amino}acetate hydrochloride

  • MDL number MFCD12197205
  • Cas Number : 1181457-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45447
Availability details for
Quantity
  • Availability
  • Add to Cart