ENA196830040 Enamine

2-(4-bromobenzamido)thiophene-3-carboxylic acid

  • MDL number MFCD09049963
  • Cas Number : 926241-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43842
Availability details for
Quantity
  • Availability
  • Add to Cart