ENA196830044 Enamine

N-(2-bromophenyl)-2-cyanoacetamide

  • MDL number MFCD03383383
  • Cas Number : 80277-87-8
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43860
Availability details for
Quantity
  • Availability
  • Add to Cart