ENA196830046 Enamine

[1-(3-chlorophenyl)ethyl](methyl)amine

  • MDL number MFCD09737462
  • Cas Number : 149529-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33189
Availability details for
Quantity
  • Availability
  • Add to Cart