ENA196830048 Enamine

2-[4-(4-bromobenzamido)phenyl]acetic acid

  • MDL number MFCD09733620
  • Cas Number : 953737-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43870
Availability details for
Quantity
  • Availability
  • Add to Cart