ENA196830051 Enamine

4-[2-(dimethylamino)ethoxy]-3-methoxybenzonitrile

  • MDL number MFCD09717755
  • Cas Number : 938368-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43877
Availability details for
Quantity
  • Availability
  • Add to Cart