ENA196830055 Enamine

7-azabicyclo[4.1.0]heptane

  • MDL number MFCD11858224
  • Cas Number : 286-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20-22
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43895

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P310,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-43895
pictograms GHS05,GHS06
hstatements H301,H314
Availability details for
Quantity
  • Availability
  • Add to Cart