ENA196830056 Enamine

2-(2-bromo-4-methylphenoxy)propanoic acid

  • MDL number MFCD08442210
  • Cas Number : 926252-65-5
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43899
Availability details for
Quantity
  • Availability
  • Add to Cart