ENA196830060 Enamine

3-{[(3-chlorophenyl)methyl]amino}propanoic acid hydrochloride

  • MDL number MFCD12197042
  • Cas Number : 1172342-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43911
Availability details for
Quantity
  • Availability
  • Add to Cart