ENA196830066 Enamine

N1-[3-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine dihydrochloride

  • MDL number MFCD12197046
  • Cas Number : 1172350-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43930
Availability details for
Quantity
  • Availability
  • Add to Cart