ENA196830067 Enamine

2-(3,3,3-trifluoropropanamido)acetic acid

  • MDL number MFCD09045159
  • Cas Number : 923972-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43939
Availability details for
Quantity
  • Availability
  • Add to Cart