ENA196830070 Enamine

1-methyl-1H-pyrazole-4-carbothioamide

  • MDL number MFCD05662461
  • Cas Number : 1152604-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43945
Availability details for
Quantity
  • Availability
  • Add to Cart