ENA196830079 Enamine

4-amino-N,N-dipropylbenzamide

  • MDL number MFCD03206821
  • Cas Number : 79868-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43975

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43975
Availability details for
Quantity
  • Availability
  • Add to Cart