ENA196830084 Enamine

2-amino-3,4-dichlorobenzoic acid

  • MDL number MFCD00078315
  • Cas Number : 20776-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-238
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43989
Availability details for
Quantity
  • Availability
  • Add to Cart