ENA196830087 Enamine

2-{[3-(ethoxycarbonyl)pyridin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD09931509
  • Cas Number : 697265-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44008
Availability details for
Quantity
  • Availability
  • Add to Cart