ENA196830092 Enamine

1-(3-methyl-1-benzofuran-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD12197056
  • Cas Number : 1172557-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44020
Availability details for
Quantity
  • Availability
  • Add to Cart