ENA196830094 Enamine

1-(furan-3-carbonyl)piperidin-4-one

  • MDL number MFCD09934412
  • Cas Number : 108206-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44026
Availability details for
Quantity
  • Availability
  • Add to Cart