ENA196830097 Enamine

2-(3,3,3-trifluoropropanamido)propanoic acid

  • MDL number MFCD09046286
  • Cas Number : 1396996-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44049
Availability details for
Quantity
  • Availability
  • Add to Cart