ENA196830099 Enamine

3-methoxy-4-(2-methoxy-2-oxoethoxy)benzoic acid

  • MDL number MFCD09935297
  • Cas Number : 94946-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44056
Availability details for
Quantity
  • Availability
  • Add to Cart