ENA196830100 Enamine

2-[3-methyl-4-(propan-2-yl)phenoxy]propanoic acid

  • MDL number MFCD02283427
  • Cas Number : 92156-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44058
Availability details for
Quantity
  • Availability
  • Add to Cart