ENA196830104 Enamine

methyl[1-(pyridin-3-yl)ethyl]amine dihydrochloride

  • MDL number MFCD12197066
  • Cas Number : 1171321-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-44074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44074
Availability details for
Quantity
  • Availability
  • Add to Cart