ENA196830106 Enamine

5-[(4-chloro-2-fluorophenyl)methyl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD12197068
  • Cas Number : 1170526-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44079
Availability details for
Quantity
  • Availability
  • Add to Cart