ENA196830111 Enamine

2-(2-fluorophenyl)-1,3-benzoxazol-5-amine

  • MDL number MFCD00579068
  • Cas Number : 313527-46-7
  • Molecular Weight : 228.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-44091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44091
Availability details for
Quantity
  • Availability
  • Add to Cart