ENA196830128 Enamine

5-(naphthalen-2-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09803846
  • Cas Number : 198893-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-252
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44155
Availability details for
Quantity
  • Availability
  • Add to Cart