ENA196830130 Enamine

3-(5-bromofuran-2-yl)-3-oxopropanenitrile

  • MDL number MFCD07785839
  • Cas Number : 93277-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40259
Availability details for
Quantity
  • Availability
  • Add to Cart