ENA196830133 Enamine

methyl 6-(benzyloxy)-1H-indole-2-carboxylate

  • MDL number MFCD04967027
  • Cas Number : 103781-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44171
Availability details for
Quantity
  • Availability
  • Add to Cart