ENA196830135 Enamine

{1-[4-(aminomethyl)phenyl]piperidin-4-yl}methanol

  • MDL number MFCD03787065
  • Cas Number : 861210-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44175

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44175
Availability details for
Quantity
  • Availability
  • Add to Cart