ENA196830145 Enamine

5-(3-chloropropyl)-1-phenyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD12197088
  • Cas Number : 73963-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44201
Availability details for
Quantity
  • Availability
  • Add to Cart