ENA196830147 Enamine

2,2,2-trifluoroethyl N-(propan-2-yl)carbamate

  • MDL number MFCD12197093
  • Cas Number : 1027616-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44207
Availability details for
Quantity
  • Availability
  • Add to Cart